Descripción
The TUBG2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TUBG2. This antibody recognizes human, mouse, rat antigen. The TUBG2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Especificaciones
Especificaciones
| Antígeno | TUBG2 |
| Aplicaciones | Immunofluorescence |
| Concentración | 1000 μg/mL |
| Conjugado | CoraLite Plus 488 |
| Símbolos de los genes | Gamma 2 tubulin, TUBG2, Tubulin gamma 2 chain, tubulin, gamma 2 |
| Especie del huésped | Rabbit |
| Cantidad | 100 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 27175 |
| Especies diana | Human, Mouse, Rat |
| Mostrar más |