missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUBA4A Monoclonal Antibody (KT204)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA549493
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This product was used in an Indirect ELISA.
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa, are homologous but not identical. The alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.
Especificaciones
| TUBA4A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P68366 | |
| TUBA4A | |
| Recombinant SARS-CoV-2 Nucleocapsid Protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| KT204 | |
| Unconjugated | |
| TUBA4A | |
| Alpha-tubulin 1; Alpha-tubulin 4; alpha-tubulin isotype M-alpha-4; ALS22; H2-ALPHA; M[a]4; Testis-specific alpha-tubulin; TUBA1; Tuba4; TUBA4A; Tubulin alpha; tubulin alpha 4a; tubulin alpha-1 chain; Tubulin alpha-4 chain; tubulin alpha-4A chain; Tubulin H2-alpha; tubulin, alpha 1 (testis specific); tubulin, alpha 4; tubulin, alpha 4a | |
| Mouse | |
| Protein A | |
| RUO | |
| 7277 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido