missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA554942
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: DCPHVDDCVF AAEFMSNDYV RVTQLYCDGV GVQYKDYIQS ERNLEFDICS IWCSKPISVL QDYCDAIKIN IFWPLLFQHQ NSSVISRLHP CVDANNSRAS EINLKKLQHL ELMEDIVDLA KKVANDS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
E3 ubiquitin-protein ligase which catalyzes the formation of 'Lys-48'-polyubiquitin chains (PubMed:20059950, PubMed:30696809). Mediates the ubiquitination and subsequent degradation of phosphorylated Akt (AKT1, AKT2 and AKT3) in the nucleus (PubMed:20059950). Acts as a terminal regulator of Akt signaling after activation; its phosphorylation by Akt, which is a prerequisite for ubiquitin ligase activity, suggests the existence of a regulation mechanism required to control Akt levels after activation (PubMed:20059950). Positively regulates TGFB1-induced epithelial-mesenchymal transition and myofibroblast differentiation by mediating the ubiquitination and subsequent degradation of SMURF2 (PubMed:30696809). Regulates neuronal differentiation by regulating actin remodeling and Golgi organization via a signaling cascade involving RHOA, CIT and ROCK (PubMed:17488780, PubMed:24695496). Inhibits cell proliferation (PubMed:30203323). [UniProt].
Especificaciones
| TTC3 | |
| Polyclonal | |
| Unconjugated | |
| TTC3 | |
| DCRR1; E3 ubiquitin-protein ligase TTC3; Protein DCRR1; RING finger protein 105; RING-type E3 ubiquitin transferase TTC3; RNF105; tetratricopeptide repeat domain 3; tetratricopeptide repeat protein 3; TPR repeat protein 3; TPR repeat protein D; TPRD; TPRDIII; TTC3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7267 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53804 | |
| TTC3 | |
| Recombinant protein corresponding to Human TTC3. Recombinant protein control fragment (Product #RP-91795). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido