missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN8 Monoclonal Antibody (657909)

Descripción
This antibody detects mouse TSPAN8 transfectants but not the parental cell line in flow cytometry. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
TSPAN8 (tetraspanin 8), also known as TM4SF3 or CO-029, is a 237 amino acid multi-pass membrane protein that belongs to the tetraspanin family of transmembrane proteins and is thought to play a role in signal transduction events that influence cell development, activation, growth and motility. Localized to the cell membrane, TSPAN8 is expressed in colon, gastric, renal and pancreatic cancers, strongly suggesting a role in cancer formation and metastasis. The gene encoding TSPAN8 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.
Especificaciones
Especificaciones
| Antígeno | TSPAN8 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Frozen) |
| Clasificación | Monoclonal |
| Clon | 657909 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 5% trehalose and No Preservative |
| génica | Tspan8 |
| N.º de referencia del gen | Q8R3G9 |
| Alias de gen | C76990; CO-029; CO-29; D6.1A; E330007O21Rik; tetraspanin 8; Tetraspanin8; tetraspanin-8; Tm4sf3; transmembrane 4 superfamily member 3; TSPAN8; Tspan-8; Tumor-associated antigen CO-029 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?