missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSLP Polyclonal Antibody

Sheep Polyclonal Antibody
Marca: Invitrogen™ PA546975
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant mouse TSLP and recombinant human TSLP R is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
Especificaciones
| TSLP | |
| Polyclonal | |
| Unconjugated | |
| TSLP | |
| Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 85480 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q969D9 | |
| TSLP | |
| E. coli-derived recombinant human TSLP Tyr29-Gln159. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido