missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSHZ2 Polyclonal Antibody

Descripción
A suggested positive control is A-20 cell lysate. PA5-34343 can be used with blocking peptide PEP-1386.
The Teashirt zinc finger homeobox (TSHZ) family comprise a family of evolutionarily conserved transcription factors that, in Drosophila, are active in specific body parts for patterning, but whose function in vertebrates is less clear. In mice, the known three TSHZ proteins are expressed in distinct patterns in the developing and adult brain, suggesting that they play a role in the establishment of regional identity and specification of cell types within the brain. Recent experiments have shown that the expression of TSHZ2 is frequently downregulated in most breast and prostate cancers and its promoter was unmethylated in virtually all cases, suggesting this family of proteins may also be involved in carcinogenesis.
Especificaciones
Especificaciones
| Antígeno | TSHZ2 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide |
| génica | TSHZ2 |
| N.º de referencia del gen | Q68FE9, Q9NRE2 |
| Alias de gen | 2900073F20Rik; B830045G17; C20orf17; cell growth-inhibiting protein 7; Kiaa4248; mKIAA4248; Mtsh2; Ovarian cancer-related protein 10-2; OVC10-2; Sdccag33l; SDCCAG33-like protein; serologically defined colon cancer antigen 33 like; teashirt 2; teashirt family zinc finger 2; teashirt homolog 2; teashirt homolog 2-like; teashirt zinc finger family member 2; teashirt zinc finger homeobox 2; teashirt2; TSH2; TSHZ2; ZABC2; Zfp218; Zinc finger protein 218; ZNF218 |
| Símbolos de los genes | TSHZ2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?