missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-56378
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TSC1 Polyclonal specifically detects TSC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones
| TSC1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| TSC1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DDYVHISLPQATVTPPRKEERMDSARPCLHRQHHLLNDRGSEEPPGSKGSVTLSDLPGFLGDLASEEDSIEKDKEEAAISRELSEITT | |
| 100 μL | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway | |
| 7248 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido