missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptose Phosphate Broth (Dehydrated)

Código de producto. p-200112225
Change view
Click to view available options
Cantidad:
500 g
5 kg
Tamaño de la unidad:
5 Kg
500 g
Código de producto. Cantidad unitSize
11983762 5 kg 5 Kg
10576143 500 g 500 g
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11983762

Marca: Thermo Scientific™ CM0283T

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Cultivate fastidious bacteria and supplement cell culture media with this medium.

Cultivate fastidious bacteria with Thermo Scientific™ Oxoid™ Tryptose Phosphate Broth (Dehydrated). Tryptose Phosphate Broth is recommended for the cultivation of streptococci, pneumococci, meningococci and other fastidious organisms. Tryptose Phosphate Broth with added agar and sodium azide is recommended for the isolation of pathogenic streptococci from cheese and other dairy products1,2.

Use Tryptone Phosphate Broth for cultivation of fastidious bacteria.

Not all products are available for sale in all territories. Please inquire

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Especificaciones

Certificaciones/conformidad Industrial Reference: FDA.
Formulario Powder
Tipo de producto Dehydrated Culture Media
Cantidad 5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.