missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPV4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA541055
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: RVDEVNWSHW NQNLGIINED PGKNETYQYY GFSHTVGRLR RDRWSSVVPR Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 100%; Rat: 93%.
TRPV4 (Transient Receptor Potential Cation Channel Subfamily V Member 4) is a cation-selective channel, activated in response to systemic osmotic pressure. Non-selective calcium permeant cation channel probably involved in osmotic sensitivity and mechanosensitivity. TRPV4 is a member of the OSM9-like transient receptor potential channel (OTRPC) subfamily in the transient receptor potential (TRP) superfamily of ion channels. The TRPV4 protein is a Ca2+-permeable, nonselective cation channel that is thought to be involved in the regulation of systemic osmotic pressure. Mutations in the TRPV4 gene are the cause of spondylometaphyseal and metatropic dysplasia and hereditary motor and sensory neuropathy type IIC. Diseases associated with TRPV4 include Metatropic Dysplasia and Spondylometaphyseal Dysplasia, Kozlowski Type. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
| TRPV4 | |
| Polyclonal | |
| Unconjugated | |
| Trpv4 | |
| 0610033B08Rik; BCYM3; CMT2C; HMSN2C; LOW QUALITY PROTEIN: transient receptor potential cation channel subfamily V member 4; OSM9-like transient receptor potential channel 4; osm-9-like TRP channel 4; osmosensitive transient receptor potential channel 4; Otrpc4; SMAL; SPSMA; SSQTL1; Transient receptor potential cation channel subfamily V member 4; transient receptor potential cation channel, subfamily 5, member 4; transient receptor potential cation channel, subfamily V, member 4; transient receptor potential protein 12; Trp12; Trpv4; trpv4 channel; Vanilloi; vanilloid receptor-like channel 2; Vanilloid receptor-like protein 2; vanilloid receptor-related osmotically activated channel; vanilloid receptor-related osmotically activated channel (Vroac); vanilloid receptor-related osmotically activated channel protein; vanilloid receptor-related osmotically-activated channel; Vrl2; VRL-2; VROAC; VR-OAC; wu:fp52e02 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 59341, 66026 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9ERZ8, Q9HBA0 | |
| Trpv4 | |
| Synthetic peptide directed towards the middle region of human TRPV4. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering