missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV1 (extracellular) Polyclonal Antibody, Invitrogen™
Descripción
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5′ UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.
Especificaciones
Especificaciones
| Antígeno | TRPV1 (extracellular) |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.8 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA and 0.05% sodium azide; pH 7.4 |
| génica | Trpv1 |
| N.º de referencia del gen | O35433 |
| Alias de gen | TRPV1 |
| Símbolos de los genes | Trpv1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?