Learn More
Invitrogen™ TRPC1 Recombinant Rabbit Monoclonal Antibody (24GB4145)
GREENER_CHOICE
Código de producto. 30276345
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30276345 100 μL 100 microlitros
30274874 20 μL 20 microlitros
2 options
Código de producto. 30276345
Marca: Invitrogen™ MA557293
Rabbit Recombinant Monoclonal Antibody
TRPC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TRPC1
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB4145
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica Trpc1
N.º de referencia del gen P48995, Q61056, Q9QX01
Alias de gen capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; Mtrp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; transient receptor protein 1; trp; TRP1; TRP-1; trpc1; trpc1.L; trp-related protein 1; Trrp1; XELAEV_18027727mg; xtrpc1
Símbolos de los genes Trpc1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human TRPC1 (aa 411-492).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 22063, 7220, 89821
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG