missing translation for 'onlineSavingsMsg'
Learn More

Troponin C (cardiac) Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30152794
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30152794 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30152794

Brand: Novus Biologicals (Bio-Techne) NBP299682CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Troponin C (cardiac) Polyclonal antibody specifically detects Troponin C (cardiac) in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Troponin C (cardiac)
Aplicaciones ELISA
Clasificación Polyclonal
Conjugado CoraFluor 1
Formulación PBS
Alias de gen cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with purified, recombinant Human Troponin C (cardiac) (Accession#: NP_003271.1; Met1-Glu161)
Método de purificación Antigen and protein A Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cell Biology, Cellular Markers
Primario o secundario Primary
ID de gen (Entrez) 7134
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.