Learn More
Invitrogen™ TrkA Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA118405
Descripción
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
Especificaciones
| TrkA | |
| Polyclonal | |
| Unconjugated | |
| Ntrk1 | |
| C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Frozen) | |
| Conc. Not Determined | |
| Whole serum with no preservative | |
| P04629, P35739, Q3UFB7 | |
| Ntrk1 | |
| Extracellular domain of glycosylated human TrkA protein produced in CHO cells. | |
| RUO | |
| 18211, 4914, 59109 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.