missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM9 Polyclonal Antibody

Descripción
Peptide sequence: DGLHFNSTYN ARVKAFNKTG VSPYSKTLVL QTSEGKALQQ YPSERELRGI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
TRIM9 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
Especificaciones
Especificaciones
| Antígeno | TRIM9 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | TRIM9 |
| N.º de referencia del gen | Q9C026 |
| Alias de gen | E3 ubiquitin-protein ligase TRIM9; fj48c11; homolog of rat RING finger Spring; hypothetical protein LOC336099; Kiaa0282; RING finger protein 91; RING-type E3 ubiquitin transferase TRIM9; RNF91; SNAP-25-interacting RING finger protein; Spring; Trim9; tripartite motif containing 9; tripartite motif protein 9; tripartite motif-containing 9; tripartite motif-containing protein 9; tripartite motif-containing protein 9; E3 ubiquitin-protein ligase TRIM9; Unknown (protein for MGC:137652); wu:fj48c11; zgc:77357 |
| Símbolos de los genes | TRIM9 |
| Mostrar más |
Título del producto
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?