missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Triadin Monoclonal Antibody (GE 4.90)

Código de producto. 11515812
Change view
Click to view available options
Cantidad:
200 μL
Tamaño de la unidad:
200 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11515812 200 μL 200 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11515812 Proveedor Invitrogen™ N.º de proveedor MA3927

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA3-927 detects triadin from mouse, rabbit and rat tissues. MA3-927 has been successfully used in Western blot, immunohistochemistry, immunoprecipitation and ELISA procedures. By Western blot, this antibody detects a 95 kDa protein representing triadin from rabbit skeletal muscle extracts. Immunohistochemical staining of triadin in rat skeletal muscle with MA3-927 results in striated staining consistent with sarcoplasmic reticulum localization. The MA3-927 antigen is purified rabbit fast skeletal muscle triadin.

The junction between the transverse tubules (T-tubules) and the sarcoplasmic reticulum (SR) of skeletal muscle is called the triad. At the triad, dihydropyridine receptors (DHPR's) of the T-tubule serve as voltage sensors in excitation-contraction coupling, while ryanodine receptors (RyR's), the calcium release channels, exist in the membrane of the terminal cisternae of the SR. It is thought that during slow phase depolarization of the T-tubule, a third protein, triadin (MW 95 kDa) transmits electrochemical signals to the SR through direct interaction with both DHPR's and RyR's. Though its exact role in this signaling process is unclear, triadin has been shown to co-localize with both DHPR and RYR at the junctional face of the terminal cisternae.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Triadin
Aplicaciones ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon GE 4.90
Concentración Conc. Not Determined
Conjugado Unconjugated
Formulación Ascites with 0.05% sodium azide
génica TRDN
N.º de referencia del gen E9Q9K5, Q28820, Q9QX75
Alias de gen 2310045H21Rik; cardiac triadin; cardiac triadin isoform 1; cardiac triadin isoform 2; cardiac triadin isoform 3; CPVT5; EG432451; junctional sarcoplasmic reticulum glycoprotein; TDN; Trdn; Triadin; triadin 1; triadin 32 kDa (TRISK 32); triadin 49 kDa (TRISK 49); triadin 95; TRISK
Símbolos de los genes TRDN
Especie del huésped Mouse
Inmunógeno Purified rabbit fast skeletal muscle triadin.
Cantidad 200 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100009519, 59299, 76757
Especies diana Mouse, Rabbit, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.