Learn More
BOLA Tri-Clamp GL-Adaptor
Descripción
- Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck.
- Universal chemical resistance, the flowing product is only exposed to PTFE.
Especificaciones
Especificaciones
| Certificaciones/conformidad | FDA |
| Esterilizables en autoclave | Autoclavable |
| Diámetro interior (métrico) | 16 mm |
| Tamaño de rosca | GL 18 |
| Color | White |