missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: RAETFVFLDL EATGLPSVEP EIAELSLFAV HRSSLENPEH DESGALVLP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
TREX2 is one of two major human 3' to 5' exonucleases which are required for checkpoint signaling after DNA damage. While the related protein TREX1 is required for ATR expression and mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis, less is known about TREX2. Like TREX1, TREX2 is ubiquitously expressed in all tissues examined. X-ray crystallography studies of TREX2-single strand DNA complexes revealed that TREX2 binds DNA as a dimer and may act to displace the second DNA strand, suggesting that TREX2 unzips and denatures double stranded DNA and feed the substrate DNA strand into the TREX2 active site. At least two isoforms of TREX2 are known to exist.
Especificaciones
Especificaciones
| Antígeno | TREX2 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.05 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | TREX2 |
| N.º de referencia del gen | Q9BQ50 |
| Alias de gen | 3'-5' exonuclease TREX2; 3'-5' exonuclease TREX2 long form; RGD1562245; Three prime repair exonuclease 2; Trex2 |
| Símbolos de los genes | TREX2 |
| Mostrar más |
Título del producto
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?
