missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREM1 Monoclonal Antibody (L5-B8)

Descripción
TREM1 Monoclonal Antibody for Western Blot, ICC/IF, Flow, Functional Assay
TREM1 (triggering receptor expressed on myeloid cells 1) is a receptor belonging to the Ig superfamily that amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. It is thought that TREM1 other related TREM proteins such as TREM2 act to modulate the inflammatory response. TREM1 has also been linked to inflammatory bowel disease resulting in enhanced production of TNF, IL-6, IL-8, and MCP-1.
Especificaciones
Especificaciones
| Antígeno | TREM1 |
| Aplicaciones | Flow Cytometry, Functional Assay, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | L5-B8 |
| Concentración | 100 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% BSA and no preservative |
| génica | TREM1 |
| N.º de referencia del gen | Q9JKE2 |
| Alias de gen | CD354; sCD354; soluble CD354; soluble TREM 1; soluble TREM1; sTREM 1; sTREM1; TREM; Trem1; TREM-1; triggering receptor expressed in monocytes 1; triggering receptor expressed on monocytes 1; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?