missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREH Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA559840
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: ALNTVLWDEQ TGAWFDYDLE KKKKNREFYP SNLTPLWAGC FSDPGVADKA LKYLEDNRIL TYQYGIPTSL QKTGQQWDFP NAWAPLQDLV IR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
This gene encodes an enzyme that hydrolyses trehalose, a disaccharide formed from two glucose molecules found mainly in fungi, plants, and insects. A partial duplication of this gene is located adjacent to this locus on chromosome 11.
Especificaciones
| TREH | |
| Polyclonal | |
| Unconjugated | |
| TREH | |
| 2210412M19Rik; Alpha,alpha-trehalase; alpha,alpha-trehalose glucohydrolase; LOW QUALITY PROTEIN: trehalase; trehalase; si:ch211-147p17.2; TRE; TREA; TREH; Trehalase; trehalase (brush-border membrane glycoprotein) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11181 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43280 | |
| TREH | |
| Recombinant protein corresponding to Human TREH. Recombinant protein control fragment (Product #RP-103880). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido