Learn More
Kartell SpA Volumetric Flasks with Screw Cap
Descripción
These flasks create no meniscus so final readings are easy. Calibrated gravimetrically at 20°C. Translucency of polypropylene enhanced when in contact with liquid. Thick walls eliminate distortion. Thin neck make final reading precise. Autoclavable. Chemically non absorbant. No wetting. Screw caps included.
Especificaciones
Especificaciones
| Capacidad (métrico) | 250 mL |
| Material | Polypropylene |
| Tipo de producto | Volumetric Flask |
| Esterilizables en autoclave | Autoclavable |
| Desechables | Reusable |
| Graduated | No |
| Mouth Style | Narrow |
| Esterilidad | Non-sterile |
| Forma del fondo | Round |
| Tratamiento de superficie | Untreated |