Learn More
transgelin/SM22 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A10C2, Proteintech
Código de producto. p-7212691 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16847225 20 μL 20 microlitros
16837225 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16847225 Proveedor Proteintech N.º de proveedor 602131IG20UL
Mouse Monoclonal Antibody
The protein encoded by this gene is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear. Two transcript variants encoding the same protein have been found for this gene.
Especificaciones
Antígeno transgelin/SM22
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon 2A10C2
Concentración 0.7 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica TAGLN
N.º de referencia del gen P31232, P37804, Q01995
Alias de gen SM22, SM22 alpha, SM22a, SM22-a, SM22alpha, Smooth muscle protein 22 alpha, TAGLN, TAGLN1, transgelin, transgelin/SM22 alpha
Símbolos de los genes TAGLN
Especie del huésped Mouse
Inmunógeno transgelin/SM22 Fusion Protein Ag0764
Método de purificación Protein G
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21345, 25123, 397021, 6876
Especies diana Rat, Human, Mouse, Pig
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1