Learn More
trans,trans-dibencilidenoacetona, 98 %, Thermo Scientific Chemicals
Tienda Thermo Scientific Chemicals ProductosDescripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Nombre del producto químico o material | trans, trans-Dibenzylideneacetone |
| Nombre de nota | 95% |
| Punto de fusión | 107°C to 114°C |
| Color | Amarillo |
| Espectro infrarrojo | Auténtico |
| Intervalo de porcentaje de ensayo | 97.5% min |
| Envase | Frasco de vidrio |
| Fórmula lineal | C6H5CH=CHC(O)CH=CHC6H5 |
| Cantidad | 5 g |
| Beilstein | 07, IV, 1747 |
| Mostrar más |