missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR2/TNFRSF10B Antibody (B-D37), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-14579AF700
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TRAILR2/TNFRSF10B Monoclonal antibody specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Flow Cytometry, ELISA
Especificaciones
| TRAILR2/TNFRSF10B | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
| Recombinant human TRAILR2/TNFRSF10B | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway | |
| 8795 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, ELISA | |
| B-D37 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido