missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (32A1380), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-27350V
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones
| TRAILR1/TNFRSF10A | |
| Monoclonal | |
| DyLight 405 | |
| O00220 | |
| TNFRSF10A | |
| This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4. | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 8797 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry | |
| 32A1380 | |
| Western Blot, Flow Cytometry | |
| APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido