missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TP53INP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marca:  Invitrogen™ PA572961

Código de producto. 15941615

  • 530.00€ / 0.10 ml

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Immunogen has 81% identity to rat. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.

TP53INP2 (tumor protein p53 inducible nuclear protein 2), also known as DOR or PINH, is a 220 amino acid nuclear protein that is expressed in developing murine brain and spinal cord, as well as in the sensory and motor neuron tracts of the peripheral nervous system. A novel protein involved in the autophagy of mammalian cells, TP53INP2 translocates from the nucleus to the autophagosome structures after activation of autophagy by rapamycin or starvation. Necessary for autophagosome development and considered a scaffold protein, TP53INP2 recruits LC3 and/or LC3-related proteins, such as GABARAP and GABARAP-like2, to the autophagosome membrane by interacting with the transmembrane protein TMEM49. The gene encoding TP53INP2 is located on human chromosome 20, which is comprised of approximately 2% of the human genome and contains nearly 63 million bases that encode over 600 genes.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

TP53INP2
Polyclonal
Unconjugated
TP53INP2
1110029F20Rik; C20orf110; diabetes and obesity regulated; diabetes and obesity-regulated; Diabetes and obesity-regulated gene; Diabetes and obesity-regulated protein; dJ1181N3.1; DOR; p53-inducible protein U; PIG-U; PINH; TP53INP2; transformation related protein 53 inducible nuclear protein 2; Trp53inp2; tumor protein p53 inducible nuclear protein 2; tumor protein p53-inducible nuclear protein 2
Rabbit
Antigen affinity chromatography
RUO
58476, 68728
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.6 mg/mL
PBS with 30% glycerol and 0.05% sodium azide
Q8CFU8, Q8IXH6
TP53INP2, Trp53inp2
A genomic peptide made to a C-terminal region of the human TP53INP2 protein (within residues 100-220).
100 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado