missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Toxoplasma gondii Polyclonal Antibody, FITC
GREENER_CHOICE

Código de producto. 11548361
Change view
Click to view available options
Cantidad:
1 mL
Tamaño de la unidad:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11548361 1 mL 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11548361 Proveedor Invitrogen™ N.º de proveedor PA17253

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

PA1-7253 detects Toxoplasma gondii. PA1-7253 has been successfully used in immunofluorescence procedures. The PA1-7253 immunogen is RH strain of Toxoplasma gondii.

Toxoplasma is a crescent shaped sporozoan that lives as an intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregnant women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregnancy, can produce blindness or mental retardation in the newborn.
TRUSTED_SUSTAINABILITY

Spécification

Antígeno Toxoplasma gondii
Aplicaciones Immunocytochemistry
Clasificación Polyclonal
Concentración 5 mg/mL
Conjugado FITC
Formulación PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Alias de gen t gondii; T. gondii
Especie del huésped Rabbit
Inmunógeno RH strain of Toxoplasma gondii.
Método de purificación Protein A
Cantidad 1 mL
Estado normativo RUO
Primario o secundario Primary
Especies diana Protozoa
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.