Learn More
Brady™ ToughStripe Max Striped Floor Tape
Click to view available options
Color:
Amarillo, negro, Black
Azul, amarillo, Yellow
Azul, blanco, White
Magenta, amarillo, Yellow
Negro, blanco, White
Negro, naranja, Orange
Rojo, amarillo, Yellow
Rojo, blanco, White
Verde, blanco, White
Anchura (métrico):
50,8 mm
76,2 mm
101,6 mm
Tamaño de la unidad:
1 rollo
Descripción
- Re-engineered to be twice as thin yet twice as durable to better withstand damaging impacts in forklift and heavy traffic areas
- Thinner profile and true edge-to-edge adhesive minimise pick points
- Strong polyester (PETG) material is four times more rigid than vinyl and contains no plasticisers
- Damage-isolating design keeps tape intact longer by preventing rips, gouges and tears from running
- Matt finish provides excellent colour contrast for clear visibility under any lighting
- Compact construction results in a smaller, lightweight roll
- Can be easily installed by one person with minimal tools
- Compatible with the ToughStripe™ Floor Tape Applicator (part number 150852)
Especificaciones
Especificaciones
| Dimensiones (L x An) | 50.8 mm x 30.48 m |
| Acabado | Gloss |
| Longitud (imperial) | 100 ft. |
| Longitud (métrico) | 30.48 m |
| Intervalo de temperatura (imperial) | 50°F to 140°F |
| Intervalo de temperatura (métrico) | 10°C to 60°C |
| Anchura (imperial) | 2 in. |
| Anchura (métrico) | 50,8 mm |
| Tipo de adhesivo | Rubber |
| Color | Amarillo, negro, Black |
| Mostrar más |