Learn More
Brady™ ToughStripe Max Striped Floor Tape
Código de producto. 30280362
Change view
Click to view available options
Color:
Amarillo, negro, Black
Azul, amarillo, Yellow
Azul, blanco, White
Magenta, amarillo, Yellow
Negro, blanco, White
Negro, naranja, Orange
Rojo, amarillo, Yellow
Rojo, blanco, White
Verde, blanco, White
Anchura (métrico):
50,8 mm
76,2 mm
101,6 mm
Tamaño de la unidad:
1 rollo
Código de producto. Color Anchura (métrico) unitSize
30280362 Verde, blanco, White 101,6 mm 1 rollo
30280380 Amarillo, negro, Black 50,8 mm 1 rollo
30280377 Amarillo, negro, Black 76,2 mm 1 rollo
30280345 Amarillo, negro, Black 101,6 mm 1 rollo
30280352 Rojo, blanco, White 50,8 mm 1 rollo
30280363 Negro, naranja, Orange 76,2 mm 1 rollo
30280353 Rojo, blanco, White 76,2 mm 1 rollo
30280381 Negro, blanco, White 50,8 mm 1 rollo
30280340 Negro, blanco, White 76,2 mm 1 rollo
30280351 Verde, blanco, White 50,8 mm 1 rollo
30280396 Rojo, blanco, White 101,6 mm 1 rollo
30280387 Negro, naranja, Orange 101,6 mm 1 rollo
30280355 Verde, blanco, White 76,2 mm 1 rollo
30280390 Azul, blanco, White 50,8 mm 1 rollo
30280395 Rojo, amarillo, Yellow 101,6 mm 1 rollo
30280343 Rojo, amarillo, Yellow 50,8 mm 1 rollo
30280357 Azul, amarillo, Yellow 101,6 mm 1 rollo
30280388 Azul, blanco, White 76,2 mm 1 rollo
30280356 Negro, naranja, Orange 50,8 mm 1 rollo
30280386 Rojo, amarillo, Yellow 76,2 mm 1 rollo
30280365 Azul, amarillo, Yellow 50,8 mm 1 rollo
30280389 Negro, blanco, White 101,6 mm 1 rollo
30280383 Azul, blanco, White 101,6 mm 1 rollo
30280369 Magenta, amarillo, Yellow 101,6 mm 1 rollo
30280392 Azul, amarillo, Yellow 76,2 mm 1 rollo
30280370 Magenta, amarillo, Yellow 76,2 mm 1 rollo
30280358 Magenta, amarillo, Yellow 50,8 mm 1 rollo
27 options
Código de producto. 30280362
Marca: Brady™ 177055
Solo quedan null
Añadir a la cesta
Añadir a la cesta
ToughStripe™ Max Floor Marking Tape is purpose-built to hold its ground in forklift and heavy traffic areas.
  • Re-engineered to be twice as thin yet twice as durable to better withstand damaging impacts in forklift and heavy traffic areas
  • Thinner profile and true edge-to-edge adhesive minimise pick points
  • Strong polyester (PETG) material is four times more rigid than vinyl and contains no plasticisers
  • Damage-isolating design keeps tape intact longer by preventing rips, gouges and tears from running
  • Matt finish provides excellent colour contrast for clear visibility under any lighting
  • Compact construction results in a smaller, lightweight roll
  • Can be easily installed by one person with minimal tools
  • Compatible with the ToughStripe™ Floor Tape Applicator (part number 150852)
Especificaciones
Dimensiones (L x An) 101.6 mm x 30.48 m
Acabado Gloss
Longitud (imperial) 100 ft.
Longitud (métrico) 30.48 m
Intervalo de temperatura (imperial) 50°F to 140°F
Intervalo de temperatura (métrico) 10°C to 60°C
Anchura (imperial) 4 in.
Anchura (métrico) 101,6 mm
Tipo de adhesivo Rubber
Color Verde, blanco, White
Material Polyester
Tipo de producto Floor Tape