missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Total Nitric Oxide and Nitrate/Nitrite Parameter Assay Kit
Descripción
The Total Nitric Oxide and Nitrate/Nitrite Parameter Assay Kit quantifies multi-species Nitric Oxide in cell culture supernates (10 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul), citrate plasma (25 ul), urine (10 ul).
Specifications
Specifications
| Intervalo del ensayo | 3.12 - 200 umol/L |
| Sensibilidad del ensayo | 0.78 umol/L |
| Conjugado | Biotina |
| Reactividad cruzada | < 50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Tipo de producto | ELISA Kit |
| Tipo de muestra | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine |
| Especificidad | Total Nitric Oxide, Nitrite and Nitrate Levels in Various Samples |
| Para utilizar con (aplicación) | ELISA |
| Cantidad | 1 kit |
| Volumen de muestra | Cell Culture Supernates (10uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL),Citrate Plasma (25uL),Urine (10uL) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?