Learn More
TOM20 Mouse anti-Human, Unconjugated, Clone: 1D6F5, Proteintech
Código de producto. 16868965 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16868965 20 μL 20 microlitros
16858965 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16868965 Proveedor Proteintech N.º de proveedor 667771IG20UL
Mouse Monoclonal Antibody
The TOM20 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TOM20. This antibody recognizes human antigen. The TOM20 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno TOM20
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 1D6F5
Concentración 536 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 9804
Especies diana Human
Isotype IgG2b