missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFAIP8L1 Polyclonal Antibody

Descripción
This target displays homology in the following species: Cow: 93%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rat: 100%.
Tumor necrosis factor, α-induced protein 8-like 1, also known as TNFα-IP 8L1, is a 186 amino acid protein belonging to the TNFα-IP 8 family. Members of the TNFα-IP 8 family are induced by nuclear factor-κB 9 (NFκB) and tumor necrosis factor (TNF), although induction by TNF is dependent upon NFκB activation. TNFα-IP 8 proteins also act as a negative mediator of apoptosis and may play a role in tumor progression. They suppress TNF-mediated apoptosis by inhibiting caspase-8 activity but not the processing of procaspase-8, subsequently resulting in inhibition of BID cleavage and caspase-3 activation.
Especificaciones
Especificaciones
| Antígeno | TNFAIP8L1 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | TNFAIP8L1 |
| N.º de referencia del gen | Q8WVP5 |
| Alias de gen | 2600017J23Rik; Oxidative stress regulated gene-beta; oxidative stress-regulated gene-beta; Oxy-beta; TIPE1; TNF alpha induced protein 8 like 1; TNF alpha-induced protein 8-like protein 1; Tnfaip8l1; TNFAIP8-like protein 1; Tumor necrosis factor alpha-induced protein 8-like protein 1; tumor necrosis factor, alpha induced protein 8 like 1; tumor necrosis factor, alpha-induced protein 8-like 1; tumor necrosis factor, alpha-induced protein 8-like protein 1 |
| Símbolos de los genes | TNFAIP8L1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?