missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tnfaip6 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16518321
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16518321 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16518321 Proveedor Invitrogen™ N.º de proveedor PA592962

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain Immunogen sequence: WGFKDGIFHN SIWLERAAGV YHREARSGKY KLTYAEAKAV CEFEGGHLAT YKQLEAARKI GFHVCAAGWM AKGRVGYPIV KPGPNCGFGK TGIIDYGIRL NRSERWDAYC YNPHAKECGG VFTDPKQIFK SPGFPNEYED NQICYWHIRL KYGQRIHLSF LDFDLEDDPG CLADYVEIYD SYDDVHGFVG RYCGDELPDD IISTGNVMTL KFLSDASVTA GGFQIKYVAM DPVSKSSQGK NTSTTSTGNK NFLAGRFSHL.

The TSG6 gene is transcribed in normal fibroblasts and activated by binding of the cytokines TNF alpha and IL-1 at AP-1 and NF-IL6 sites in its promoter. TSG-6 is a glycoprotein and a member of the hyaluronan-binding protein family, which includes cartilage link protein, proteoglycan core protein and the adhesion receptor CD44. TSG-6 is highly homologous to CD44, particularly in the hyaluronic acid-binding domain. TSG-6 is found in TNF-treated cells; its expression is rapidly activated by TNF alpha, IL-1 and lipopolysaccharide in normal fibroblasts, peripheral blood mononuclear cells, synovial cells and chondrocytes. The presence of TSG-6 in synovial fluid suggests a possible role in rheumatoid arthritis. TSG-6 forms a stable complex with components of the serine protease inhibitor, inter-alpha-inhibitor (I alpha I)). TSG-6 potentiates the inhibitory effect of l alpha l on the protease activity of plasmin. Through their cooperative inhibitory effect on plasmin, TSG-6 and l alpha l can modulate the protease network and thus inhibit inflammation.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Tnfaip6
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 1.32 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
génica TNFAIP6
N.º de referencia del gen O08859, P98066
Alias de gen Hyaluronate-binding protein; TNF alpha induced protein 6; TNF alpha-induced protein 6; Tnfaip6; Tnfip6; TNF-stimulated gene 6 protein; TSG6; TSG-6; tumor necrosis factor alpha induced protein 6; tumor necrosis factor alpha-induced protein 6; tumor necrosis factor alpha-inducible protein 6; tumor necrosis factor induced protein 6; tumor necrosis factor, alpha induced protein 6; tumor necrosis factor, alpha-induced protein 6; tumor necrosis factor-inducible gene 6 protein; tumor necrosis factor-stimulated gene-6 protein
Símbolos de los genes TNFAIP6
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 18-277 of human TNFAIP6 (NP_0090462).
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21930, 7130, 84397
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.