missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF beta Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13111-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.Especificaciones
| TNF beta | |
| Polyclonal | |
| Unconjugated | |
| LTA | |
| LT, LT alpha, LTA, Lymphotoxin alpha, TNF beta, TNFB, TNFSF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4049 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P01374 | |
| LTA | |
| TNF beta Fusion Protein Ag3775 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido