missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNF beta Polyclonal Antibody
GREENER_CHOICE

Código de producto. 30535742
Change view
Click to view available options
Cantidad:
1 mg
Tamaño de la unidad:
1 miligramo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30535742 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30535742 Proveedor Invitrogen™ N.º de proveedor PA5148441

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

Reconstitute at 1 mg/mL in sterile PBS.

TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TNF beta
Aplicaciones Neutralization, Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with trehalose and no preservative
génica LTA
N.º de referencia del gen P01374
Alias de gen DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF β; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFβ; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1E; tumor necrosis factor ligand superfamily member 1
Símbolos de los genes LTA
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant human Lymphotoxin-alpha /TNF-beta, Leu35-Leu205.
Método de purificación Protein A/G
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4049
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.