missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS11D Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA542876
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: IHSVCLPAAT QNIPPGSTAY VTGWGAQEYA GHTVPELRQG QVRIISNDVC Sequence homology: Human: 100%.
The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
Especificaciones
| TMPRSS11D | |
| Polyclonal | |
| Unconjugated | |
| TMPRSS11D | |
| Adrenal secretory serine protease; airway spesific trypsin-like protease; airway trypsin like protease; airway trypsin-like protease; AsP; AST; AT; BC020151; HAT; Mat; MGC150587; MGC150588; Rat; TMPRSS11D; Transmembrane protease serine 11D; Transmembrane protease serine 11D catalytic chain; Transmembrane protease serine 11D non-catalytic chain; transmembrane protease, serine 11D; transmembrane serine protease 11D | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9407 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O60235 | |
| TMPRSS11D | |
| Synthetic peptide directed towards the middle region of human TMPRSS11D. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido