Learn More
Invitrogen™ TMF1 Recombinant Rabbit Monoclonal Antibody (24GB1965)
Descripción
TMF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
Especificaciones
Especificaciones
| Antígeno | TMF1 |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 24GB1965 |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | TMF1 |
| N.º de referencia del gen | P82094 |
| Alias de gen | 7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1 |
| Símbolos de los genes | TMF1 |
| Mostrar más |