Learn More
TMEM97 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 16844905 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16844905 20 μL 20 microlitros
16834905 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16844905 Proveedor Proteintech N.º de proveedor 264441AP20UL
Rabbit Polyclonal Antibody
The TMEM97 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMEM97. This antibody recognizes human, mouse antigen. The TMEM97 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
Antígeno TMEM97
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Polyclonal
Concentración 300 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 27346
Especies diana Human, Mouse
Isotype IgG