Learn More
TMEM70 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16859924 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16859924 20 μL 20 microlitros
16849924 150 μL 150 microlitros
2 options
Código de producto. 16859924
Marca: Proteintech 203881AP20UL
Rabbit Polyclonal Antibody
TMEM70 is a recently identified mitochondrial protein that is thought to play a role in the biogenesis of the ATP synthase in higher eukaryotes. Mutations in this gene result in early neonatal onset of hypotonia, hypertrophic cardiomyopathy, lactic acidos and 3-methylglutaconic aciduria (3-MGC-uria), and usually cause death within the first six weeks of life, although some patients survive much longer. Little is known of the role of TMEM70, but it is conserved across multicellular eukaryotic organisms. It contains a conserved DUF1301 domain and two putative transmembrane regions.
Especificaciones
Antígeno TMEM70
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.26 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica TMEM70
N.º de referencia del gen Q9BUB7
Alias de gen TMEM70, transmembrane protein 70
Símbolos de los genes TMEM70
Especie del huésped Rabbit
Inmunógeno TMEM70 Fusion Protein Ag14226
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 54968
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG