missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM204 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15721061 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15721061 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15721061

Marca: Invitrogen™ PA553112

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: HKREDCMAPR VIVISRSLTA RFRRGLDNDY VESPC Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Transmembrane protein 204 (TMEM204) is a four-transmembrane protein that is induced under hypoxic conditions. It localizes to cell-cell junctions and co-localizes with the beta-catenin adherens junction-associated protein, but not with tight junctions. Overexpression of TMEM204 results in decreased adhesion between cells, indicating that TMEM204 is an adherens junction protein that modulates cell adhesion and potentially angiogenesis. Recent evidence suggests TMEM204 may also be involved in epilepsy.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TMEM204
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica TMEM204
N.º de referencia del gen Q9BSN7
Alias de gen C16orf30; Claudin-like protein 24; claudin-like protein of 24 kDa; Clp24; Tmem204; Transmembrane protein 204; UNQ6509/PRO21434
Símbolos de los genes TMEM204
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human TMEM204. Recombinant protein control fragment (Product #RP-90756).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79652
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.