Learn More
TMEM173/STING Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16859334 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16859334 20 μL 20 microlitros
16849334 150 μL 150 microlitros
2 options
Código de producto. 16859334
Marca: Proteintech 198511AP20UL
Rabbit Polyclonal Antibody
STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.
Especificaciones
Antígeno TMEM173/STING
Aplicaciones Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Clasificación Polyclonal
Concentración 0.33 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica TMEM173
N.º de referencia del gen F1M391, Q3TBT3, Q86WV6
Alias de gen hMITA, hSTING, Mediator of IRF3 activation, MITA, MPYS, NET23, STING, TMEM173, transmembrane protein 173
Símbolos de los genes TMEM173
Especie del huésped Rabbit
Inmunógeno TMEM173/STING Fusion Protein Ag13921
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 340061, 498840, 72512
Especies diana Rat, Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG