missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM119 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17742104
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
17742104 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17742104

Marca: Invitrogen™ PA5120705

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive test controls include: Mouse liver, Mouse brain, Rat brain. Immunogen sequence: SRPEEALDSS RQLQADILAA TQNLKSPTRA ALGGGDGARM VEGRGAEEEE KGSQEGDQEV QGHGVPVETP EAQEEPCSGV LEGAVVAGEG QGELEGSLLL AQEAQGPVGP P.

Tmem119 encodes a single-pass type I transmembrane protein which seems to interact with a multitude of micro-RNAs as well as certain transcription factors such as SMAD1, SMAD5 and RUNX2. Tmem119 has been identified as a surface marker of microglia that can be used to reliably distinguish microglia from infiltrating macrophages. Microglial cells are phagocytes that are restricted to the brain and involved in clearing up debris and dendrite pruning of neurons. They display a high functional plasticity, balancing neuroprotective and neurotoxic functions, which is reflected by their distinct physiology in response to changes in their microenvironment. Microglia are particularly mobile in areas of the brain undergoing restructuring. Reactive microglia co-express the marker Iba1. In a pathological context, microglia have been associated with differentiation of pathogenic/inflammatory A1 type astrocytes by secretion of Cq1, IL-1a and TNF when stimulated with LPS, and thought to contribute to the pathology of several neurodegenerative diseases such as Alzheimers, Parkinsons, Chorea Huntingtons disease and Multiple Sclerosis. The specific function Tmem119 plays on microglial cells remains unclear. Outside of the neural system, Tmem119 seems to be involved in osteoblast differentiation/ossification.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TMEM119
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 2.11 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
génica TMEM119
N.º de referencia del gen Q8R138
Alias de gen AW208946; BC025600; OBIF; Osteoblast induction factor; PSEC0199; Tmem119; Transmembrane protein 119; UNQ731/PRO1415
Símbolos de los genes TMEM119
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 160-270 of human TMEM119 (NP_859075.2).
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 231633, 304581
Especies diana Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.