missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMC7 Polyclonal Antibody

Descripción
Predicted to react with Mouse and Rat samples.
TMC7 (Transmembrane channel-like protein 7) is a 723 amino acid protein that is a member of the TMC protein family. All TMC genes encode transmembrane proteins with intracellular amino- and carboxy- termini and at least eight membrane spanning domains. Therefore, TMC7 is a multi-pass membrane protein that may regulate or function as an ion channel or transporter. The gene encoding TMC7 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
Especificaciones
Especificaciones
| Antígeno | TMC7 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.4 |
| génica | TMC7 |
| N.º de referencia del gen | Q7Z402, Q8C428 |
| Alias de gen | 1700030H01Rik; C230064B05; C630024K23Rik; RGD1564267; Tmc7; transmembrane channel like 7; transmembrane channel-like 7; transmembrane channel-like gene family 7; transmembrane channel-like protein 7 |
| Símbolos de los genes | TMC7 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?