missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TM9SF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA534269
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey (100%) Marmoset, Elephant, Dog, Bat, Bovine, Horse, Rabbit, Pig (93%).
TM9SF4 is a protein coding gene.
Especificaciones
| TM9SF4 | |
| Polyclonal | |
| Unconjugated | |
| TM9SF4 | |
| AA986553; AU045326; B930079E06; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 4; dJ836N17.2; Kiaa0255; LOW QUALITY PROTEIN: transmembrane 9 superfamily member 4; mKIAA0255; TM9SF4; Transmembrane 9 superfamily member 4; transmembrane 9 superfamily protein member 4; TUCAP1; Tumor cannibalism associated protein 1; zgc:66234 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9777 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q92544 | |
| TM9SF4 | |
| Synthetic 15 amino acid peptide from internal region of human TM9SF4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido