missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Marca: Novus Biologicals NBP2-24827SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TLR5 Polyclonal specifically detects TLR5 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, In vitro assay, Flow (Cell Surface).
Especificaciones
| TLR5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human, Mouse and Rat. Porcine reactivity reported in scientific literature (PMID: 24097563). | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), In vitro Assay | |
| 1.0 mg/mL | |
| Western Blot 1 - 3 ug/mL, Flow Cytometry 1 - 2 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 30716659), Immunohistochemistry-Paraffin, In vitro assay reported in scientific literature (PMID 26084721), Flow (Cell Surface) reported in scientific literature (PMID 24532644) | |
| O60602 | |
| TLR5 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 found between amino acids 300-350. | |
| 0.025 mg | |
| Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
| 7100 | |
| Human, Mouse, Rat, Pig | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido