Descripción
The TLR4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse TLR4. This antibody recognizes mouse, human antigen. The TLR4 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | TLR4 |
| Aplicaciones | Western Blot, ELISA |
| Concentración | 550 μg/mL |
| Símbolos de los genes | Lps, Ly87, Ran/M1, Rasl2 8, Tlr4, toll like receptor 4 |
| Especie del huésped | Rabbit |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 21898 |
| Especies diana | Mouse, Human |
| Contenido y almacenamiento | Store at -20°C. Stable for one year after shipment. |
| Mostrar más |