missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-56388-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TLE1 Polyclonal specifically detects TLE1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones
| TLE1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| TLE1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TSATPGLRPGLGKPPAIDPLVNQAAAGLR | |
| 25 μL | |
| Signal Transduction | |
| 7088 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido