Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Tienda Epredia Productos
Click to view available options
Color:
Aguamarina
Amarillo
Azul
Blanco
Canela
Gris
Lila
Naranja
Rosa
Verde
Tamaño de la unidad:
paquete de 1000
paquete de 500
Descripción
- Saves time, offers the greatest specimen security, and provides ease of use
- Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
- Eliminates the need for messy sponges or lens paper
- Ensures tissues are safely processed as cassettes will not pop open during processing
Especificaciones
Especificaciones
| Tipo de poro | Mesh Pore Insert |
| Color | Lila |
| Tipo de producto | Casete HistoScreen™ |