Learn More
Tissue alpha-L-Fucosidase/FUCA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Especificaciones
Antígeno Tissue alpha-L-Fucosidase/FUCA1
Dilución Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 uL/mg of lysate
Aplicaciones Western Blot, ELISA, Immunoprecipitation
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18648306
Ver documentos
Novus Biologicals
NBP3-00209-100ul
100 μg
326.00€
100 microlitros
Ver productos alternativos
Ahorre hasta 
»
18637136
Ver documentos
Novus Biologicals
NBP3-00209-50ul
50 μg
216.00€
50 microlitros
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
Tissue alpha-L-Fucosidase/FUCA1 Polyclonal antibody specifically detects Tissue alpha-L-Fucosidase/FUCA1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Especificaciones
Especificaciones
Tissue alpha-L-Fucosidase/FUCA1
Western Blot, ELISA, Immunoprecipitation
Unconjugated
Rabbit
Signal Transduction
0.2 um filtered solution in PBS
2517
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 uL/mg of lysate
Polyclonal
Purified
RUO
Human
Alpha-L-fucosidase 1, Alpha-L-fucosidase I, Alpha-L-fucoside fucohydrolase 1, EC 3.2.1, EC 3.2.1.51, FUCA, fucosidase, alpha-L- 1, tissue, tissue alpha-L-fucosidase
Produced in rabbits immunized with purified, recombinant Human Tissue alpha-L-Fucosidase/FUCA1 (Accession#: P04066; Gln32-Lys466)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.