Learn More
TIPE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15940-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Acts as a negative regulator of innate and adaptive immunity by maintaining immune homeostasis. Negative regulator of Toll-like receptor and T-cell receptor function. Prevents hyperresponsiveness of the immune system and maintains immune homeostasis. Inhibits JUN/AP1 and NF-kappa-B activation. Promotes Fas-induced apoptosis (By similarity).Especificaciones
| TIPE2 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP8L2 | |
| Inflammation factor protein 20, TIPE2, TNFAIP8 like protein 2, TNFAIP8L2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 310663, 69769, 79626 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AYJ8, Q6P589, Q9D8Y7 | |
| Tnfaip8l2 | |
| TIPE2 Fusion Protein Ag7964 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |