missing translation for 'onlineSavingsMsg'
Learn More

TIMP4, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Código de producto. 16000758
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16000758 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16000758 Supplier Abnova Supplier No. PAB8015.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specifications

Antígeno TIMP4
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulación Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO4, 0.05 mg Thimerosal, 0.05 mg NaN3.
génica TIMP metallopeptidase inhibitor 4
Alias de gen -
Símbolos de los genes TIMP4
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to amino acids at C-terminus of human TIMP4.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7079
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.